1-(3-(Difluoromethoxy)phenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65481-500MG 65481-1G 65481-5G
77336-926EA 400.67 USD
77336-926 77356-840 77380-812
1-(3-(Difluoromethoxy)phenyl)-2,2,2-trifluoroethanone ≥95%
1-(3-(Difluoromethoxy)phenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅F₅O₂
MW: 240.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22688451
CAS Number: 1417459-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR