1-(2-Chloro-5-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65471-500MG 65471-1G 65471-25G 65471-5G
77332-554EA 307.97 USD
77332-554 77356-834 77389-816 77380-808
1-(2-Chloro-5-methoxyphenyl)-2,2,2-trifluoroethanone ≥95%
1-(2-Chloro-5-methoxyphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₂
MW: 238.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28105091
CAS Number: 1824063-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR