1-(4-Chloro-3,5-dimethylphenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65464-500MG 65464-1G 65464-5G 65464-25G
77332-550EA 131.84 USD
77332-550 77344-376 77368-602 77389-814
1-(4-Chloro-3,5-dimethylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(4-Chloro-3,5-dimethylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₈ClF₃O
MW: 236.618 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113541
CAS Number: 1256482-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR