1-(4-Chloro-2,5-dimethylphenyl)-2,2,2-trifluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
65462-500MG
65462-1G
65462-5G
65462-25G
77332-546EA
161.71
USD
77332-546
77345-950
77368-598
77389-810
1-(4-Chloro-2,5-dimethylphenyl)-2,2,2-trifluoroethanone ≥97%
1-(4-Chloro-2,5-dimethylphenyl)-2,2,2-trifluoroethanone
Precaución: for Research Use Only
Formula:
C₁₀H₈ClF₃O Storage Temperature: Ambient |
MDL Number:
MFCD24107309 CAS Number: 1507684-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...