3,5-Dibromo-4-(2,2,2-trifluoroethoxy)pyridine ≥95%

Supplier: AOBChem USA

Warning

65450-500MG 65450-1G 65450-5G 65450-25G
77332-540EA 120.51 USD
77332-540 77343-788 77368-592 77380-802
3,5-Dibromo-4-(2,2,2-trifluoroethoxy)pyridine ≥95%
3,5-Dibromo-4-(2,2,2-trifluoroethoxy)pyridine

Precaución: for Research Use Only

Formula: C₇H₄Br₂F₃NO
MW: 334.916 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762225
CAS Number: 1357094-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR