3,5-Dibromo-4-(cyclopropylmethoxy)pyridine ≥95%

Supplier: AOBChem USA

Warning

65448-500MG 65448-1G 65448-5G 65448-25G
77332-536EA 99.91 USD
77332-536 77342-862 77368-588 77380-798
3,5-Dibromo-4-(cyclopropylmethoxy)pyridine ≥95%
3,5-Dibromo-4-(cyclopropylmethoxy)pyridine

Precaución: for Research Use Only

Formula: C₉H₉Br₂NO
Storage Temperature: Ambient
MDL Number: MFCD34762223
CAS Number: 2807446-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR