4-(Benzyloxy)-3,5-dibromopyridine ≥95%

Supplier: AOBChem USA

Warning

65447-500MG 65447-1G 65447-5G 65447-25G
77332-534EA 99.91 USD
77332-534 77342-860 77368-586 77380-796
4-(Benzyloxy)-3,5-dibromopyridine ≥95%
4-(Benzyloxy)-3,5-dibromopyridine

Precaución: for Research Use Only

Formula: C₁₂H₉Br₂NO
MW: 343.014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762222
CAS Number: 2807454-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR