3-Bromo-5-iodo-4-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

65441-500MG 65441-1G 65441-5G 65441-25G
77332-530EA 157.59 USD
77332-530 77345-658 77368-582 77389-802
3-Bromo-5-iodo-4-methoxypyridine ≥97%
3-Bromo-5-iodo-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrINO
MW: 313.918 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762220
CAS Number: 2807454-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR