4,4,5,5-Tetramethyl-2-(2,3,6-trifluorophenyl)-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

65431-1G 65431-5G 65431-25G
77340-798EA 60.77 USD
77340-798 77368-580 77380-794
4,4,5,5-Tetramethyl-2-(2,3,6-trifluorophenyl)-1,3,2-dioxaborolane ≥97%
4,4,5,5-Tetramethyl-2-(2,3,6-trifluorophenyl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₂H₁₄BF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD24039525
CAS Number: 1819398-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR