(4-Methylpiperazin-1-yl)(2,3,6-trifluorophenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

65430-1G 65430-5G 65430-25G
77340-906EA 62.83 USD
77340-906 77368-578 77380-792
(4-Methylpiperazin-1-yl)(2,3,6-trifluorophenyl)methanone ≥97%
(4-Methylpiperazin-1-yl)(2,3,6-trifluorophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD34762212
CAS Number: 2918899-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR