Piperidin-1-yl(2,3,6-trifluorophenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

65427-1G 65427-5G 65427-25G
77341-356EA 82.4 USD
77341-356 77368-572 77380-786
Piperidin-1-yl(2,3,6-trifluorophenyl)methanone ≥97%
Piperidin-1-yl(2,3,6-trifluorophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₂F₃NO
Storage Temperature: Ambient
MDL Number: MFCD34762209
CAS Number: 2807470-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR