Pyrrolidin-1-yl(2,3,6-trifluorophenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

65426-1G 65426-5G 65426-25G
77341-354EA 82.4 USD
77341-354 77368-570 77380-784
Pyrrolidin-1-yl(2,3,6-trifluorophenyl)methanone ≥97%
Pyrrolidin-1-yl(2,3,6-trifluorophenyl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₀F₃NO
MW: 229.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762208
CAS Number: 2285556-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR