2,3,6-Trifluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

65425-1G 65425-5G 65425-25G
77341-090EA 70.04 USD
77341-090 77368-568 77380-782
2,3,6-Trifluoro-N-methoxy-N-methylbenzamide ≥97%
2,3,6-Trifluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD34762207
CAS Number: 2289149-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR