2,3,6-Trifluoro-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

65424-1G 65424-5G 65424-25G
77341-576EA 93.73 USD
77341-576 77368-566 77380-780
2,3,6-Trifluoro-N-methylbenzamide ≥97%
2,3,6-Trifluoro-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₆F₃NO
MW: 189.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30289558
CAS Number: 1516695-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR