2,4,4',5-Tetrafluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

65419-500MG 65419-1G 65419-5G 65419-25G
77332-528EA 182.31 USD
77332-528 77346-998 77368-564 77389-800
2,4,4',5-Tetrafluoro-1,1'-biphenyl ≥97%
2,4,4',5-Tetrafluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₆F₄
MW: 226.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762206
CAS Number: 2901084-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR