4'-Bromo-2,4,5-trifluoro-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

65418-500MG 65418-1G 65418-5G 65418-25G
77332-526EA 182.31 USD
77332-526 77346-996 77368-562 77389-798
4'-Bromo-2,4,5-trifluoro-1,1'-biphenyl ≥95%
4'-Bromo-2,4,5-trifluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₆BrF₃
MW: 287.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762205
CAS Number: 2901098-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR