2,4,5-Trifluoro-4'-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

65415-500MG 65415-1G 65415-5G 65415-25G
77332-524EA 141.11 USD
77332-524 77344-864 77368-560 77389-796
2,4,5-Trifluoro-4'-methyl-1,1'-biphenyl ≥95%
2,4,5-Trifluoro-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉F₃
MW: 222.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762202
CAS Number: 1246661-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR