2,6-Difluoro-3-(2,2,2-trifluoroacetyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

65370-500MG 65370-1G 65370-5G 65370-25G
77332-518EA 161.71 USD
77332-518 77345-948 77368-554 77389-792
2,6-Difluoro-3-(2,2,2-trifluoroacetyl)benzonitrile ≥97%
2,6-Difluoro-3-(2,2,2-trifluoroacetyl)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₂F₅NO
Storage Temperature: Ambient
MDL Number: MFCD34762201
CAS Number: 1536911-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR