(2-Fluoro-4-methylphenyl)(thiomorpholino)methanone ≥95%

Supplier: AOBChem USA

Warning

65342-500MG 65342-1G 65342-5G 65342-25G
77332-514EA 161.71 USD
77332-514 77345-946 77368-550 77389-788
(2-Fluoro-4-methylphenyl)(thiomorpholino)methanone ≥95%
(2-Fluoro-4-methylphenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄FNOS
MW: 239.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23768629
CAS Number: 1540739-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR