(2-Fluoro-4-methylphenyl)(piperidin-1-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

65340-500MG 65340-1G 65340-5G 65340-25G
77332-510EA 108.15 USD
77332-510 77343-196 77368-546 77380-776
(2-Fluoro-4-methylphenyl)(piperidin-1-yl)methanone ≥95%
(2-Fluoro-4-methylphenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆FNO
MW: 221.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22731608
CAS Number: 1511648-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR