(2-Fluoro-4-methylphenyl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

65339-1G 65339-5G 65339-25G
77341-886EA 106.09 USD
77341-886 77368-544 77380-774
(2-Fluoro-4-methylphenyl)(pyrrolidin-1-yl)methanone ≥97%
(2-Fluoro-4-methylphenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄FNO
MW: 207.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23344978
CAS Number: 1516371-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR