N-Cyclohexyl-2-fluoro-4-methylbenzamide ≥95%

Supplier: AOBChem USA

Warning

65337-500MG 65337-1G 65337-5G 65337-25G
77332-508EA 99.91 USD
77332-508 77342-858 77368-540 77380-770
N-Cyclohexyl-2-fluoro-4-methylbenzamide ≥95%
N-Cyclohexyl-2-fluoro-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO
Storage Temperature: Ambient
MDL Number: MFCD24765871
CAS Number: 1508439-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR