4'-Fluoro-2,3-dimethyl-1,1'-biphenyl ≥97%
Supplier: AOBChem USA
|
Warning
|
65332-500MG
65332-1G
65332-5G
65332-25G
77332-504EA
116.39
USD
77332-504
77343-448
77368-536
77380-766
4'-Fluoro-2,3-dimethyl-1,1'-biphenyl ≥97%
4'-Fluoro-2,3-dimethyl-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₄H₁₃F Storage Temperature: Ambient |
MDL Number:
MFCD34762196 CAS Number: 72968-90-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...