4'-Methoxy-2,3-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

65331-500MG 65331-1G 65331-5G 65331-25G
77332-502EA 233.81 USD
77332-502 77356-824 77368-534 77389-784
4'-Methoxy-2,3-dimethyl-1,1'-biphenyl ≥97%
4'-Methoxy-2,3-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₆O
MW: 212.287 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762195
CAS Number: 1427675-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR