2,3,4'-Trimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

65330-500MG 65330-1G 65330-5G 65330-25G
77332-500EA 161.71 USD
77332-500 77345-942 77368-532 77389-782
2,3,4'-Trimethyl-1,1'-biphenyl ≥97%
2,3,4'-Trimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₆
MW: 196.288 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762194
CAS Number: 76708-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR