4-Bromo-4'-methoxy-2,3-dimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

65325-500MG 65325-1G 65325-5G 65325-25G
77332-496EA 161.71 USD
77332-496 77345-940 77368-528 77389-778
4-Bromo-4'-methoxy-2,3-dimethyl-1,1'-biphenyl ≥95%
4-Bromo-4'-methoxy-2,3-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO
MW: 291.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34641917
CAS Number: 2807449-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR