4-Bromo-4'-fluoro-2,3-dimethyl-1,1'-biphenyl ≥95%
Supplier: AOBChem USA
|
Warning
|
65324-500MG
65324-1G
65324-5G
65324-25G
77332-494EA
178.19
USD
77332-494
77346-670
77368-526
77389-776
4-Bromo-4'-fluoro-2,3-dimethyl-1,1'-biphenyl ≥95%
4-Bromo-4'-fluoro-2,3-dimethyl-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₄H₁₂BrF Storage Temperature: Ambient |
MDL Number:
MFCD34762193 CAS Number: 2807465-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...