2-Chloro-4-fluoro-5-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

65309-500MG 65309-1G 65309-5G
77339-666EA 612.85 USD
77339-666 77356-816 77380-760
2-Chloro-4-fluoro-5-formylbenzoic acid ≥97%
2-Chloro-4-fluoro-5-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClFO₃
MW: 202.567 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762188
CAS Number: 2386463-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR