2-Chloro-4-fluoro-5-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

65307-500MG 65307-1G 65307-5G
77336-650EA 390.37 USD
77336-650 77356-814 77380-758
2-Chloro-4-fluoro-5-iodobenzaldehyde ≥97%
2-Chloro-4-fluoro-5-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClFIO
MW: 284.454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30180145
CAS Number: 2092282-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR