4-Chloro-2-fluoro-5-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

65296-500MG 65296-1G 65296-5G
77335-890EA 367.71 USD
77335-890 77356-812 77380-756
4-Chloro-2-fluoro-5-formylbenzoic acid ≥97%
4-Chloro-2-fluoro-5-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClFO₃
MW: 202.567 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762185
CAS Number: 2385984-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR