4-Bromo-2-chloro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

65274-500MG 65274-1G 65274-5G
77338-810EA 519.12 USD
77338-810 77356-806 77380-750
4-Bromo-2-chloro-5-methylbenzaldehyde ≥97%
4-Bromo-2-chloro-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrClO
MW: 233.4896 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22566293
CAS Number: 1416351-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR