(4-Bromo-2-chloro-5-methylphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

65270-500MG 65270-1G 65270-25G 65270-5G
77332-490EA 290.46 USD
77332-490 77356-804 77389-772 77380-748
(4-Bromo-2-chloro-5-methylphenyl)boronic acid ≥95%
(4-Bromo-2-chloro-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrClO₂
MW: 249.2972 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24449166
CAS Number: 2225155-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR