tert-Butyl (4-bromo-2,5-difluorophenyl)carbamate ≥97%

Supplier: AOBChem USA

Warning

65265-500MG 65265-1G 65265-5G 65265-25G
77332-488EA 232.78 USD
77332-488 77356-800 77368-522 77389-770
tert-Butyl (4-bromo-2,5-difluorophenyl)carbamate ≥97%
tert-Butyl (4-bromo-2,5-difluorophenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrF₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD34762171
CAS Number: 1874418-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR