(3-Methyl-[1,1'-biphenyl]-4-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

65256-500MG 65256-1G 65256-5G 65256-25G
77332-486EA 181.28 USD
77332-486 77346-892 77368-520 77389-768
(3-Methyl-[1,1'-biphenyl]-4-yl)methanol ≥95%
(3-Methyl-[1,1'-biphenyl]-4-yl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O
MW: 198.2603 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27920898
CAS Number: 1028202-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR