N-(4-Bromo-2,5-difluorophenyl)-2,2,2-trifluoroacetamide ≥97%
Supplier: AOBChem USA
|
Warning
|
65251-500MG
65251-1G
65251-5G
65251-25G
77332-482EA
161.71
USD
77332-482
77345-938
77368-518
77389-766
N-(4-Bromo-2,5-difluorophenyl)-2,2,2-trifluoroacetamide ≥97%
N-(4-Bromo-2,5-difluorophenyl)-2,2,2-trifluoroacetamide
Precaución: for Research Use Only
Formula:
C₈H₃BrF₅NO MW: 304.0115 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34762167 CAS Number: 1878666-40-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...