2-Fluoro-3-(trimethylsilyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

65225-500MG 65225-1G 65225-5G 65225-25G
77332-480EA 264.71 USD
77332-480 77356-796 77368-516 77389-764
2-Fluoro-3-(trimethylsilyl)benzaldehyde ≥97%
2-Fluoro-3-(trimethylsilyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₃FOSi
MW: 196.2935 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762156
CAS Number: 2807447-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR