2,2,2-Trifluoro-1-(2-fluoro-5-(trifluoromethyl)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

65218-500MG 65218-1G 65218-5G 65218-25G
77332-478EA 184.37 USD
77332-478 77347-272 77368-514 77389-762
2,2,2-Trifluoro-1-(2-fluoro-5-(trifluoromethyl)phenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(2-fluoro-5-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₃F₇O
MW: 260.1083 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762150
CAS Number: 1517587-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR