1-(4-Bromo-3-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65216-500MG 65216-1G 65216-5G 65216-25G
77332-476EA 272.95 USD
77332-476 77356-790 77368-512 77389-760
1-(4-Bromo-3-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(4-Bromo-3-chloro-2-fluorophenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₈H₂BrClF₄O
MW: 305.4514 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29959938
CAS Number: 1695359-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR