1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65210-500MG 65210-1G 65210-25G 65210-5G
77332-474EA 299.73 USD
77332-474 77356-788 77389-758 77380-738
1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone ≥95%
1-(6-Bromo-2-fluoro-3-(methylthio)phenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅BrF₄OS
MW: 317.0980 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762148
CAS Number: 2807466-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR