1-(2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

65202-500MG 65202-1G 65202-5G 65202-25G
77332-468EA 161.71 USD
77332-468 77345-936 77368-508 77389-754
1-(2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥95%
1-(2-Bromo-6-fluoro-4-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₂BrF₇O
MW: 339.0044 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762145
CAS Number: 2807444-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR