1-(2-Bromo-3-fluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65197-500MG 65197-1G 65197-5G 65197-25G
77332-466EA 184.37 USD
77332-466 77347-270 77368-506 77389-752
1-(2-Bromo-3-fluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%
1-(2-Bromo-3-fluoropyridin-4-yl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₇H₂BrF₄NO
Storage Temperature: Ambient
MDL Number: MFCD34762144
CAS Number: 2807470-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR