1-(2-Chloro-5-fluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

65195-500MG 65195-1G 65195-25G 65195-5G
77332-464EA 305.91 USD
77332-464 77356-782 77389-750 77380-734
1-(2-Chloro-5-fluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%
1-(2-Chloro-5-fluoropyridin-4-yl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₇H₂ClF₄NO
Storage Temperature: Ambient
MDL Number: MFCD34762142
CAS Number: 2807448-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR