(6-Bromo-3-chloro-2-fluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

65192-1G 65192-5G 65192-25G
77341-050EA 66.95 USD
77341-050 77368-502 77380-732
(6-Bromo-3-chloro-2-fluorophenyl)trimethylsilane ≥97%
(6-Bromo-3-chloro-2-fluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₁BrClFSi
MW: 281.6245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762140
CAS Number: 2807447-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR