(3-Bromo-6-chloro-2-fluorophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

65191-500MG 65191-1G 65191-5G 65191-25G
77332-460EA 100.94 USD
77332-460 77342-960 77368-500 77380-730
(3-Bromo-6-chloro-2-fluorophenyl)trimethylsilane ≥97%
(3-Bromo-6-chloro-2-fluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₁BrClFSi
Storage Temperature: Ambient
MDL Number: MFCD29914190
CAS Number: 2166921-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR