(3-Bromo-6-chloro-2-fluoro-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

65183-500MG 65183-1G 65183-5G 65183-25G
77332-456EA 120.51 USD
77332-456 77343-784 77368-496 77380-726
(3-Bromo-6-chloro-2-fluoro-5-methylphenyl)(methyl)sulfane ≥97%
(3-Bromo-6-chloro-2-fluoro-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₇BrClFS
MW: 269.5615 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762138
CAS Number: 2807440-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR