3-Bromo-6-chloro-2-fluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

65178-500MG 65178-1G 65178-5G 65178-25G
77332-454EA 149.35 USD
77332-454 77345-320 77368-494 77389-746
3-Bromo-6-chloro-2-fluoro-5-methylbenzoic acid ≥97%
3-Bromo-6-chloro-2-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD31637749
CAS Number: 1820035-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR