Methyl 5-chloro-6-fluoropicolinate ≥97%
Supplier: AOBChem USA
|
Warning
|
65166-500MG
65166-1G
65166-5G
77337-252EA
409.94
USD
77337-252
77356-778
77380-722
Methyl 5-chloro-6-fluoropicolinate ≥97%
Methyl 5-chloro-6-fluoropicolinate
Precaución: for Research Use Only
Formula:
C₇H₅ClFNO₂ MW: 189.5715 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14698140 CAS Number: 1214335-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...