2-Fluoro-6-iodo-3-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

65146-500MG 65146-1G
77340-544EA 1711.86 USD
77340-544 77356-774
2-Fluoro-6-iodo-3-methylpyridine ≥97%
2-Fluoro-6-iodo-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅FIN
Storage Temperature: Ambient
CAS Number: 1803765-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR