N-(4-Bromo-2-fluoro-5-methylphenyl)-2,2,2-trifluoroacetamide ≥97%

Supplier: AOBChem USA

Warning

65139-500MG 65139-1G 65139-5G 65139-25G
77332-448EA 182.31 USD
77332-448 77346-994 77368-490 77389-742
N-(4-Bromo-2-fluoro-5-methylphenyl)-2,2,2-trifluoroacetamide ≥97%
N-(4-Bromo-2-fluoro-5-methylphenyl)-2,2,2-trifluoroacetamide

Precaución: for Research Use Only

Formula: C₉H₆BrF₄NO
MW: 300.0476 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34762118
CAS Number: 1877091-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR