N-(4-Bromo-2-fluoro-5-methylphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

65138-500MG 65138-1G 65138-5G 65138-25G
77332-446EA 106.09 USD
77332-446 77343-136 77368-488 77380-718
N-(4-Bromo-2-fluoro-5-methylphenyl)acetamide ≥97%
N-(4-Bromo-2-fluoro-5-methylphenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₉BrFNO
MW: 246.0762 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30176160
CAS Number: 1864304-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR